missing translation for 'onlineSavingsMsg'
Learn More

CYB5R3 Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10894-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837583

  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CYB5R3 (NADH-cytochrome b5, reductase 3, diaphorase 1) is a 301 amino acid protein encoded by the human gene CYB5R3. CYB5R3 belongs to the flavoprotein pyridine nucleotide cytochrome reductase family and has two naturally occurring isoforms. Isoform 1 is anchored to the cytoplasmic side of the endoplasmic reticulum membrane and mitochondrion outer membrane, while isoform 2 is the soluble form found in erythrocytes. CYB5R3 is involved in the desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism and, in erythrocytes, methemoglobin reduction.
Spécification

Spécification

CYB5R3
Polyclonal
Unconjugated
CYB5R3
B5R, CYB5R3, Cytochrome b5 reductase, cytochrome b5 reductase 3, DIA1, Diaphorase 1, NADH cytochrome b5 reductase 3
Rabbit
Antigen Affinity Chromatography
RUO
100524254, 109754, 1727, 25035
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.18 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P00387, P20070, Q9DCN2
CYB5R3, LOC100524254
CYB5R3 Fusion Protein Ag1339
150 μL
Primary
Rat, Porcine, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions