missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP1A1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13241-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene, CYP1A1, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by some polycyclic aromatic hydrocarbons (PAHs), some of which are found in cigarette smoke. The enzyme′s endogenous substrate is unknown; however, it is able to metabolize some PAHs to carcinogenic intermediates. The gene has been associated with lung cancer risk. A related family member, CYP1A2, is located approximately 25 kb away from CYP1A1 on chromosome 15.Spécification
CYP1A1 | |
Polyclonal | |
Unconjugated | |
CYP1A1 | |
AHH, AHRR, CP11, CYP1, CYP1A1, CYPIA1, Cytochrome P450 1A1, Cytochrome P450 C, Cytochrome P450 form 6, Cytochrome P450 P1, P1 450, P450 C, P450DX | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13076, 1543 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00184, P04798 | |
Cyp1a1 | |
CYP1A1 Fusion Protein Ag3990 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |