missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDAH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14966-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity.Spécification
DDAH2 | |
Polyclonal | |
Unconjugated | |
DDAH2 | |
DDAH, DDAH 2, DDAH2, DDAHII, Dimethylargininase 2, G6a, NG30, Protein G6a, S phase protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23564, 294239, 51793 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95865, Q6MG60, Q99LD8 | |
Ddah2 | |
DDAH2 Fusion Protein Ag6831 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |