missing translation for 'onlineSavingsMsg'
Learn More

DHX29 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13923-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16832354

  • 121.38€ / 20µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

DNA helicase and probable main scaffold component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair; according to PubMed:20687897 the contribution to DNA double-strand break repair appears to be largely indirect through transcriptional regulation. Recruited by YY1 to YY1-activated genes, where it acts as an essential coactivator. Binds DNA. In vitro, has double-stranded DNA-dependent ATPase activity. Involved in UV-damage excision repair, DNA replication and chromosome segregation during normal cell division cycle.
Spécification

Spécification

DHX29
Polyclonal
Unconjugated
DHX29
DDX29, DEAH box protein 29, DHX29, Nucleic acid helicase DDXx
Rabbit
Antigen Affinity Chromatography
RUO
100362324, 218629, 54505
-20°C
Liquid
Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6PGC1, Q7Z478
DHX29
DHX29 Fusion Protein Ag4938
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions