missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLEU1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11860-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
May act as a tumor suppressor.Spécification
DLEU1 | |
Polyclonal | |
Unconjugated | |
DLEU1 | |
DLB1, DLEU1, DLEU2, LEU1, LEU2, NCRNA00021, XTP6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10301 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43261 | |
DLEU1 | |
DLEU1 Fusion Protein Ag2435 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |