missing translation for 'onlineSavingsMsg'
Learn More

DNAJB8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 17071-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16836904

  • 121.38€ / 20µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

DNAJB8 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain.
Spécification

Spécification

DNAJB8
Polyclonal
Unconjugated
DNAJB8
DJ6, DNAJB8
Rabbit
Antigen Affinity Chromatography
RUO
165721, 500253, 56691
-20°C
Liquid
Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NHS0, Q9QYI7
DNAJB8
DNAJB8 Fusion Protein Ag10899
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions