missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13853-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
DNAJC4 | |
Polyclonal | |
Unconjugated | |
DNAJC4 | |
DANJC4, DnaJ like protein HSPF2, DNAJC4, HSPF2, MCG18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3338, 361717, 57431 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9D844, Q9NNZ3 | |
DNAJC4 | |
DNAJC4 Fusion Protein Ag4826 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |