missing translation for 'onlineSavingsMsg'
Learn More

DNAL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 15809-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16835164

  • 121.38€ / 20µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

DNAL1 was identified as a potential candidate gene for primary ciliary dyskinesia (PCD), a genetically heterologous disorder characterized by chronic infections of the upper and lower airways that often leads to permanent lung damage, randomization of left/right body symmetry, and reduced fertility. DNAL1 is reported to be expressed solely in tissues carrying motile cilia for flagella and interacts with DNAH5, a protein that when mutated has been shown to result in PCD. It has been suggested that DNAL1 serves a regulatory function for DNAH5 activity in outer dynein arms of sperm flagella, respiratory cilia, and ependymal cilia. DNAL1 has also been recently identified as an HIV dependency factor (HDF), suggesting that DNAL1 may be an important drug target in HIV treatment. At least two isoforms of DNAL1 are known to exist.
Spécification

Spécification

DNAL1
Polyclonal
Unconjugated
DNAL1
C14orf168, DNAL1, Dynein light chain 1, axonemal
Rabbit
Antigen Affinity Chromatography
RUO
105000, 685664, 83544
-20°C
Liquid
Western Blot, Immunoprecipitation
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q05A62, Q4LDG9
Dnal1
DNAL1 Fusion Protein Ag8545
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions