Learn More
Invitrogen™ DPP2 Recombinant Rabbit Monoclonal Antibody (003), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA543457
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Specificity: Human DPP7/DPPII/DPP2
The protein encoded by this gene is a post-proline cleaving aminopeptidase expressed in quiescent lymphocytes. The resting lymphocytes are maintained through suppression of apoptosis, a state which is disrupted by inhibition of this novel serine protease. The enzyme has strong sequence homology with prolylcarboxypeptidase and is active at both acidic and neutral pH.
Spécification
| DPP2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9UHL4 | |
| DPP7 | |
| Recombinant Human DPP7/DPPII/DPP2 Protein. | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 003 | |
| Unconjugated | |
| DPP7 | |
| carboxytripeptidase; dipeptidyl aminopeptidase II; dipeptidyl arylamidase II; dipeptidyl peptidase 2; Dipeptidyl peptidase 7; dipeptidyl peptidase II; dipeptidyl-peptidase 2; dipeptidylpeptidase 7; dipeptidyl-peptidase II; DPP II; DPP2; Dpp7; DPPII; QPP; Quiescent cell proline dipeptidase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 29952 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |