Learn More
Invitrogen™ DPP2 Recombinant Rabbit Monoclonal Antibody (003), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
256.00€ - 512.00€
Spécification
| Antigène | DPP2 |
|---|---|
| Clone | 003 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
17759803
|
Invitrogen™
MA543457 |
50 μL |
256.00€
50µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
17799633
![]() |
Invitrogen™
MA543085 |
100 μL |
512.00€
100µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Specificity: Human DPP7/DPPII/DPP2
The protein encoded by this gene is a post-proline cleaving aminopeptidase expressed in quiescent lymphocytes. The resting lymphocytes are maintained through suppression of apoptosis, a state which is disrupted by inhibition of this novel serine protease. The enzyme has strong sequence homology with prolylcarboxypeptidase and is active at both acidic and neutral pH.Spécification
| DPP2 | |
| 1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| carboxytripeptidase; dipeptidyl aminopeptidase II; dipeptidyl arylamidase II; dipeptidyl peptidase 2; Dipeptidyl peptidase 7; dipeptidyl peptidase II; dipeptidyl-peptidase 2; dipeptidylpeptidase 7; dipeptidyl-peptidase II; DPP II; DPP2; Dpp7; DPPII; QPP; Quiescent cell proline dipeptidase | |
| DPP7 | |
| IgG | |
| Protein A | |
| Antibody |
| 003 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9UHL4 | |
| 29952 | |
| Recombinant Human DPP7/DPPII/DPP2 Protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| DPP7 |