Learn More
DRG2 Mouse anti-Human, Unconjugated, Clone: 3A3E1, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67164-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16829645
  • 121.38€ / 20µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 
Description
Description
The DRG2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DRG2. This antibody recognizes human, mouse, rat, pig antigen. The DRG2 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
DRG2
Monoclonal
1000 μg/mL
Mouse
Primary
Pig, Rat, Mouse, Human
ELISA, Immunofluorescence, Western Blot
3A3E1
Unconjugated
20 μL
1819
IgG2a