Learn More
DUOX1 Mouse anti-Human, Unconjugated, Clone: 3B8D6, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67226-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16829755
  • 121.38€ / 20µL
Expédition estimée: 03-10-2022
Description
The DUOX1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DUOX1. This antibody recognizes human antigen. The DUOX1 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
DUOX1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
ELISA, Immunofluorescence, Western Blot
3B8D6
Unconjugated
20 μL
53905
IgG2a