missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYRK2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP1-76560-0.025mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
DYRK2 Polyclonal antibody specifically detects DYRK2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Spécification
| DYRK2 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| dual specificity tyrosine-phosphorylation-regulated kinase 2, dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2, EC 2.7.12, EC 2.7.12.1, FLJ21217, FLJ21365 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human DYRK2. The immunogen is located within amino acids 520 - 570 of DYRK2. Amino Acid Squence: LRRRLPKPPTGEKTSCKR | |
| 0.025 mg | |
| Apoptosis, Protein Kinase | |
| 8445 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 10-20 μg/mL | |
| NP_006473 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu