missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E2F7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24489-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
E2F transcription factors, such as E2F7, play an essential role in the regulation of cell cycle progression.Spécification
E2F7 | |
Polyclonal | |
Unconjugated | |
E2F7 | |
E2F transcription factor 7, E2F7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
144455 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96AV8 | |
E2F7 | |
E2F7 Fusion Protein Ag21599 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |