missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EnquireBio Elastin (ELN) (Marker of Arterial Stiffness and Atherosclerosis) Mouse, Clone: ELN/1981, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 2006-MSM1-P1
Informations supplémentaires : Poids : 0.01000kg
Spécification
Elastin (ELN) (Marker of Arterial Stiffness and Atherosclerosis) | |
Secreted and Extracellular Space | |
ELN/1981 | |
Unconjugated | |
Mouse | |
70kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
2006 | |
IgG1 κ |
Immunohistochemistry, ELISA | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human Elastin protein | |
A549 or U-2197cells. Liver, Kidney, Lung or Intestine. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |