missing translation for 'onlineSavingsMsg'
Learn More

EML1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 12765-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16830664

  • 121.38€ / 20µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Human echinoderm microtubule-associated protein-like is a strong candidate for the Usher syndrome type 1A gene. Usher syndromes (USHs) are a group of genetic disorders consisting of congenital deafness, retinitis pigmentosa, and vestibular dysfunction of variable onset and severity depending on the genetic type. The disease process in USHs involves the entire brain and is not limited to the posterior fossa or auditory and visual systems. The USHs are categorized as type I (USH1A, USH1B, USH1C, USH1D, USH1E and USH1F), type II (USH2A and USH2B) and type III (USH3). The type I is the most severe form. Gene loci responsible for these three types are all mapped. Two transcript variants encoding different isoforms have been found for this gene.
Spécification

Spécification

EML1
Polyclonal
Unconjugated
EML1
ELP79, EMAP, EMAP 1, EMAP1, EMAPL, EMAPL1, EML1, HuEMAP, HuEMAP 1
Rabbit
Antigen Affinity Chromatography
RUO
2009, 362783, 68519
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00423, Q05BC3, Q4V8C3
Eml1
EML1 Fusion Protein Ag3429
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions