missing translation for 'onlineSavingsMsg'
Learn More

ENO3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 55234-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16836725

  • 325.38€ / 150µL
Expédition estimée: 04-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is specific to ENO3. It has no cross reaction to ENO1 and ENO2.

This gene encodes one of the three enolase isoenzymes found in mammals. This isoenzyme is found in skeletal muscle cells in the adult where it may play a role in muscle development and regeneration. A switch from alpha enolase to beta enolase occurs in muscle tissue during development in rodents. Mutations in this gene have be associated glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been described.
Spécification

Spécification

ENO3
Polyclonal
Unconjugated
ENO3
Beta enolase, ENO3, ENO3-specific, Enolase 3, enolase 3 (beta, muscle), MSE, Muscle specific enolase, Skeletal muscle enolase
Rabbit
Antigen Affinity Chromatography
RUO
13808, 2027, 25438
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.76 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P13929, P15429, P21550
Eno3
Peptide
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions