Learn More
Entactin Mouse anti-Human, Unconjugated, Clone: 2A11G1, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67310-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The Entactin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Entactin. This antibody recognizes human, pig, mouse, rat antigen. The Entactin antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Entactin | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, ELISA | |
2A11G1 | |
Unconjugated | |
20 μL | |
4811 | |
IgG1 |