Learn More
Entactin Mouse anti-Human, Unconjugated, Clone: 2A11G1, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67310-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16829865
  • 140.00€ / 20µL
Expédition estimée: 15-05-2024
Description
The Entactin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Entactin. This antibody recognizes human, pig, mouse, rat antigen. The Entactin antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
Entactin
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, ELISA
2A11G1
Unconjugated
20 μL
4811
IgG1