missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENTPD6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11626-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
ENTPD6 | |
Polyclonal | |
Unconjugated | |
ENTPD6 | |
CD39 antigen like 2, CD39L2, ENTPD6, NTPDase 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12497, 85260, 955 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75354, Q9ER31 | |
Entpd6 | |
ENTPD6 Fusion Protein Ag2148 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |