missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Evi2a Rabbit anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 51014-2-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
May complex with itself or/and other proteins within the membrane, to function as part of a cell-surface receptor.Spécification
Evi2a | |
Polyclonal | |
Unconjugated | |
Evi2a | |
AW491894, Evi 2, Evi2, Evi2a | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14017 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P20934 | |
Evi2a | |
Evi2a Fusion Protein Ag0478 | |
150 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |