Learn More
F11R Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16183-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The F11R antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human F11R. This antibody recognizes human, mouse antigen. The F11R antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Spécification
F11R | |
Polyclonal | |
Unconjugated | |
20 μL | |
50848 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
133 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |