Learn More
F11R Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 16183-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16835724
  • 140.00€ / 20µL
Expédition estimée: 06-05-2024
Description
The F11R antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human F11R. This antibody recognizes human, mouse antigen. The F11R antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Spécification
Spécification
F11R
Polyclonal
Unconjugated
20 μL
50848
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA
133 μg/mL
Rabbit
Primary
Human, Mouse