Learn More
F4/80 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28463-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The F4/80 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse F4/80. This antibody recognizes mouse antigen. The F4/80 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Spécification
| F4/80 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 13733 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
| 367 μg/mL | |
| Rabbit | |
| Primary | |
| Mouse |