missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP2/I-FABP Antibody (SR1211), Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP3-22293-100ul
This item is not returnable.
View return policy
Description
FABP2/I-FABP Monoclonal antibody specifically detects FABP2/I-FABP in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| FABP2/I-FABP | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1211 | |
| Western Blot 1:500-1:2000 | |
| FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein | |
| A synthesized peptide derived from human FABP2/I-FABP (Uniprot #: P12104) | |
| 100 μg | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 2169 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction