missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP3 Mouse anti-Human, Clone: 2B5C1, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 60280-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Heart-type fatty acid binding protein (hFABP), also known as mammary-derived growth inhibitor, is a protein that in humans is encoded by the FABP3 gene. The intracellular fatty acid-binding proteins (FABPs) belong to a multigene family. Fatty acid-binding protein 3 gene contains four exons and its function is to arrest growth of mammary epithelial cells. This gene is also a candidate tumor suppressor gene for human breast cancer. Cardiac-type fatty acid-binding protein (cFABP) from human heart muscle of three individuals was isolated and characterized as pI 5.3-cFABP.Spécification
FABP3 | |
Monoclonal | |
1.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05413 | |
FABP3 | |
FABP3 Fusion Protein Ag21483 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot | |
2B5C1 | |
Unconjugated | |
FABP3 | |
FABP11, FABP3, Fatty acid binding protein 3, H FABP, M FABP, MDGI, O FABP | |
Mouse | |
Protein A | |
RUO | |
2170 | |
-20°C | |
Liquid |