missing translation for 'onlineSavingsMsg'
Learn More

FAM120A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 21529-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16831125

  • 121.38€ / 20µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

53BP1 (P53-binding protein 1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, 53BP1 is phosphorylated and localizes to the presumptive sites of damage, specifically, double-strand breaks. 53BP1 may have a role in checkpoint signaling during mitosis, enhances TP53-mediated transcriptional activation, and participates in DNA repair by maintaining genomic stability.
Spécification

Spécification

FAM120A
Polyclonal
Unconjugated
FAM120A
C9orf10, DNAPTP1, DNAPTP5, FAM120A, KIAA0183, OSSA, Protein FAM120A
Rabbit
Antigen Affinity Chromatography
RUO
218236, 23196, 291019
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6A0A9, Q9NZB2
Fam120a
FAM120A Fusion Protein Ag16149
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions