missing translation for 'onlineSavingsMsg'
Learn More

FAM120C Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 25987-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16834515

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a potential transmembrane protein and lies in a region where mutations and deletions have been associated with intellectual disability and autism. Alternative splicing results in multiple transcript variants.
Spécification

Spécification

FAM120C
Polyclonal
Unconjugated
FAM120C
CXorf17, FAM120C, ORF34, Protein FAM120C, Tumor antigen BJ HCC 21
Rabbit
Antigen Affinity Chromatography
RUO
54954
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NX05
FAM120C
FAM120C Fusion Protein Ag23235
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions