missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM55B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27877-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
FAM55B | |
Polyclonal | |
Unconjugated | |
NXPE2 | |
FAM55B, FAM55B Ens | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
120406 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96DL1 | |
NXPE2 | |
FAM55B Fusion Protein Ag27351 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |