missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fatty acid desaturase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP1-84311-25ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Fatty acid desaturase 2 Polyclonal specifically detects Fatty acid desaturase 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Spécification
| Fatty acid desaturase 2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| D6DLLCDL2, Delta(6) desaturase, Delta(6) fatty acid desaturase, Delta-6 desaturase, delta-6 fatty acid desaturase, delta-6-desaturase, DES6, EC 1.14.19, EC 1.14.19.-, FADSD6, fatty acid desaturase 2, linoleoyl-CoA desaturase (delta-6-desaturase)-like 2, SLL0262, TU13 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9415 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FADS2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GKGGNQGEGAAEREVSVPTFSWEEIQKHNLRTDRWLVIDRKVYNITKWSIQHPGGQRVIGHYAGEDATDAFRAFHPDLEFVGKFLKPLLIGELAPEEPSQDHGKNSKITEDFRALRKTAEDMNLFKTNH | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu