missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO44 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10626-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
F-box proteins, a critical component of the evolutionary conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Novel human F-box proteins named FBG1, FBG2, FBG3, FBG4 and FBG5 constitute a third subfamily of mammalian F-box proteins. FBG genes are expressed in a limited number of human tissues including kidney, liver, brain and muscle tissues.Spécification
FBXO44 | |
Polyclonal | |
Unconjugated | |
FBXO44 | |
F box only protein 44, F box protein 44, F box protein FBX30, F box/G domain protein 3, FBG3, FBX30, Fbx44, FBX6A, FBXO44, Fbxo6a | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230903, 500587, 93611 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BK26, Q9H4M3 | |
Fbxo44 | |
FBXO44 Fusion Protein Ag0977 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |