missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FcεR1α Mouse anti-Human, BV510, Clone: AER-37 (also known as CRA-1, CRA1), BD OptiBuild™
Mouse Monoclonal Antibody
Marque: BD Biosciences 747786
Informations supplémentaires : Poids : 0.01370kg
Description
FcεR1α Monoclonal antibody specifically detects FcεR1α in Human samples. It is validated for Flow Cytometry.Spécification
FcεR1α | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
AER-37 (also known as CRA-1, CRA1) | |
Brilliant Violet 510 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2b κ |