missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRLA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26949-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Receptors for the Fc fragment of IgG, or FCGRs, are cell surface glycoproteins of the Ig superfamily (IgSF). These receptors mediate phagocytosis of IgG-coated pathogens and promote activation of effector cells, leading to inflammatory responses and antibody-mediated cellular cytotoxicity. All FCGR genes map to human chromosome 1. Additional genes in this region, including FREB, encode FCGR homologs that are selectively expressed in B cells and may be implicated in B-cell development and lymphomagenesis.Spécification
FCRLA | |
Polyclonal | |
Unconjugated | |
FCRLA | |
Fc receptor like A, Fc receptor like protein, Fc receptor related protein X, FCRL, FCRL1, FCRLA, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRLM1, FCRLX, FCRX, FREB, RP11 474I16.5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84824 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7L513 | |
FCRLA | |
FCRLA Fusion Protein Ag25331 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |