missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FDPS Recombinant Rabbit Monoclonal Antibody (HL2123), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen MA550388
Informations supplémentaires : Poids : 0.01000kg
Description
The isoprene biosynthetic pathway supply the cell with cholesterol, ubiquinone, and various nonsterol metabolites. The farnesylpyrophosphate synthetase enzyme catalyzes the formation of geranyl and farnesylpyrophosphate from isopentenylpyrophosphate and dimethylallyl pyrophosphate. Analysis of FDPS activity and protein in rat liver, accompanied by immunofluorescence and immunoelectron microscopy studies, demonstrated that FDPS is predominantly localized in peroxisomes. 1 Liver tissue from patients with the peroxisomal deficiency diseases Zellweger syndrome and neonatal adrenoleukodystrophy exhibit diminished activities of FDPS and subsequent isoprenoid synthesis.Spécification
FDPS | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P05369, P14324, Q920E5 | |
FDPS | |
Recombinant fragment of human FDPS. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
HL2123 | |
Unconjugated | |
FDPS | |
(2E,6E)-farnesyl diphosphate synthase, 6030492I17Rik, Ac2-125, AI256750, cholesterol-regulated 39 kDa protein, CR 39, dimethylallyltranstransferase, Farnesyl diphosphate synthase, farnesyl diphosphate synthase (farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase), farnesyl diphosphate synthetase, farnesyl pyrophosphate synthase, farnesyl pyrophosphate synthetase, farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase, Fdps, Fdpsl1, FPP synthase, FPP synthetase, FPPS, FPS, geranyltranstransferase, KIAA1293, mKIAA1293, POROK9, testis-specific farnesyl pyrophosphate synthetase | |
Rabbit | |
Protein A | |
RUO | |
110196, 2224, 83791 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |