missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FDX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12592-1-AP-20UL
Additional Details : Poids : 0.00010kg
Description
The product of this gene is a small iron-sulfur protein that transfers electrons from NADPH through ferredoxin reductase to a terminal cytochrome P450. This particular oxidation/reduction system is found in steroidogenic tissues, and is involved with the synthesis of bile acid and vitamin D. In addition to the expressed gene at this chromosomal locus (11q22), there are pseudogenes located on chromosomes 20 and 21. This gene product has been identified in a number of different tissues but all forms have been shown to be identical and are not tissue specific.Specifications
FDX1 | |
Polyclonal | |
Unconjugated | |
FDX1 | |
Adrenal ferredoxin, Adrenodoxin, mitochondrial, ADX, FDX, FDX1, ferredoxin 1, Hepatoredoxin, LOH11CR1D | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14148, 2230, 29189 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P10109, P24483, P46656 | |
Fdx1 | |
FDX1 Fusion Protein Ag3301 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |