missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FDX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | FDX1 |
---|---|
Concentration | 0.53 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16830384
|
Proteintech
12592-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 11-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16820384
|
Proteintech
12592-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 11-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The product of this gene is a small iron-sulfur protein that transfers electrons from NADPH through ferredoxin reductase to a terminal cytochrome P450. This particular oxidation/reduction system is found in steroidogenic tissues, and is involved with the synthesis of bile acid and vitamin D. In addition to the expressed gene at this chromosomal locus (11q22), there are pseudogenes located on chromosomes 20 and 21. This gene product has been identified in a number of different tissues but all forms have been shown to be identical and are not tissue specific.Spécification
FDX1 | |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P10109, P24483, P46656 | |
14148, 2230, 29189 | |
FDX1 Fusion Protein Ag3301 | |
Primary | |
-20°C | |
FDX1 |
0.53 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Adrenal ferredoxin, Adrenodoxin, mitochondrial, ADX, FDX, FDX1, ferredoxin 1, Hepatoredoxin, LOH11CR1D | |
Fdx1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |