missing translation for 'onlineSavingsMsg'
Learn More

FGF13 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 26235-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16894745

  • 349.00€ / 150µL
Expédition estimée: 16-12-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. This gene is located in a region on chromosome X, which is associated with Borjeson-Forssman-Lehmann syndrome (BFLS), making it a possible candidate gene for familial cases of the BFLS, and for other syndromal and nonspecific forms of X-linked mental retardation mapping to this region. Alternative splicing of this gene at the 5′ end results in several transcript variants encoding different isoforms with different N-termini.
Spécification

Spécification

FGF13
Polyclonal
Unconjugated
FGF13
FGF 13, FGF13, FGF2, FHF 2, FHF2, fibroblast growth factor 13
Rabbit
Antigen Affinity Chromatography
RUO
14168, 2258
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P70377, Q92913
FGF13
FGF13 Fusion Protein Ag23965
150 μL
Primary
Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions