missing translation for 'onlineSavingsMsg'
Learn More

FGF17 Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

Marque:  Invitrogen 500-P152-1MG

Informations supplémentaires : Poids : 0.16500kg

 Afficher plus de versions de ce produit

Code produit. 17894593

  • 1596.00€ / 1mg
Expédition estimée: 25-10-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

AA Sequence of recombinant protein: MTQGENHPSP NFNQYVRDQG AMTDQLSRRQ IREYQLYSRT SGKHVQVTGR RISATAEDGN KFAKLIVETD TFGSRVRIKG AESEKYICMN KRGKLIGKPS GKSKDCVFTE IVLENNYTAF QNARHEGWFM AFTRQGRPRQ ASRSRQNQRE AHFIKRLYQG QLPFPNHAEK QKQFEFVGSA PTRRTKRTRR PQPLT. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human FGF-17. Anti-Human FGF-17-specific antibody was purified by affinity chromatography employing an immobilized Human FGF-17 matrix. Sandwich ELISA: To detect Human FGF-17 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human FGF-17 (500-P152Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human FGF-17. Western Blot: To detect Human FGF-17 by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human FGF-17 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

FGF17 Polyclonal antibody specifically detects FGF17 in Human samples. It is validated for ELISA, Western Blot
Spécification

Spécification

FGF17
Polyclonal
PBS with no preservative
O60258
Rabbit
Antigen affinity chromatography
RUO
8822
-20°C
ELISA, Western Blot
Unconjugated
FGF17
FGF; FGF-13; FGF17; FGF-17; Fibroblast growth factor; Fibroblast growth factor 17; HH20; UNQ161/PRO187
E.coli-derived Recombinant Human FGF-17
1 mg
Primary
Human
Lyophilized
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions