missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF21 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26272-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.Spécification
FGF21 | |
Polyclonal | |
Unconjugated | |
FGF21 | |
FGF 21, FGF21, fibroblast growth factor 21 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26291 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NSA1 | |
FGF21 | |
FGF21 Fusion Protein Ag24064 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |