missing translation for 'onlineSavingsMsg'
Learn More

FH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10966-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837703

  • 325.38€ / 150µL
Expédition estimée: 10-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a protein that is localized in the mitochondrial inner compartment and that may play a role in mitochondrial apoptosis. Nonsense mutations have been reported to result in cytochrome c oxidase deficiency.
Spécification

Spécification

FH
Polyclonal
Unconjugated
FH
FH, Fumarase, fumarate hydratase, HLRCC, LRCC, MCL, MCUL1
Rabbit
Antigen Affinity Chromatography
RUO
14193, 2271, 24368
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Flow Cytometry
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P07954, P14408, P97807
FH
FH Fusion Protein Ag1437
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions