missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13582-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
FKBP6 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP6 is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin.This gene encodes a member of the interferon regulatory transcription factor (IRF) family. Family members share a highly-conserved N-terminal helix-turn-helix DNA-binding domain and a less conserved C-terminal protein-binding domain. Mutations in this gene can cause van der Woude syndrome and popliteal pterygium syndrome. This protein is involved in palate formation.Spécification
FKBP6 | |
Polyclonal | |
Unconjugated | |
FKBP6 | |
36 kDa FK506 binding protein, 36 kDa FKBP, FK506 binding protein 6, FK506 binding protein 6, 36kDa, FKBP 36, FKBP 6, FKBP36, FKBP6, Immunophilin FKBP36, PPIase, PPIase FKBP6, Rotamase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
288597, 8468, 94244 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3ZQF4, O75344, Q91XW8 | |
Fkbp6 | |
FKBP6 Fusion Protein Ag4505 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |