Learn More
FLNC Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 28492-1-AP-20UL
Informations supplémentaires : Poids : 0.00020kg
 Afficher plus de versions de ce produit
Code produit. 17271406
  • 129.00€ / 20µL
Expédition estimée: 14-12-2022
pour voir le stock disponible 
Description
Description
Filamin C (FLNC; also known as γ-FLN; ABP-L; FLN2), a muscle-specific filamin and a large actin-cross-linking protein. Human filamins are ∼280 kDa homodimers, each monomer consisting of an N-terminal actin-binding domain followed by 24 immunoglobulin (Ig)-like domains (d1 to d24), the last of which mediates dimerization. FLNC is specifically expressed in cardiomyocytes and skeletal myocytes and is involved in the maintenance of structural integrity. High filamin C associated with better prognosis of prostate cancer, leukemia and breast cancer patients. ( PMID: 25577646, PMID: 30867563, PMID: 31131323)
Spécification
Spécification
FLNC
Polyclonal
Unconjugated
20 μL
2318
Liquid
ELISA, Western Blot
300 μg/mL