Learn More
FOXA2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 22474-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16891945
  • 129.00€ / 20µL
Expédition estimée: 16-12-2022
Description
FOXA2 is a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific genes such as albumin and transthyretin. They also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. FOXA2 has been linked to sporadic cases of maturity-onset diabetes of the young. Transcript variants encoding different isoforms have been identified.
Spécification
Spécification
FOXA2
Polyclonal
Unconjugated
FOXA2
forkhead box A2, Forkhead box protein A2, FOXA2, HNF 3 beta, HNF 3B, HNF3B, TCF 3B, TCF3B, Transcription factor 3B
Rabbit
Antigen Affinity Chromatography
RUO
3170
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y261
FOXA2
FOXA2 Fusion Protein Ag18221