missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxO1/FKHR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-24857SS
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
FoxO1/FKHR Polyclonal specifically detects FoxO1/FKHR in Human samples. It is validated for Western Blot.
Spécification
| FoxO1/FKHR | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat and bovine, 83% homologous in zebrafish and pufferfish, and 77% homologous in chicken. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1.0 mg/mL | |
| Western Blot 0.05-0.50 ug/ml | |
| Q12778 | |
| FOXO1 | |
| A portion of amino acids 250-300 of human FOXO1A protein was used as the immunogen. | |
| 0.025 mg | |
| Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers | |
| 2308 | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu