Learn More
GAD1 / GAD67 (GABAergic Neuronal Marker) Mouse, Clone: GAD1/2391, enQuireBio™
Mouse Monoclonal Antibody
Marque:  EnquireBio 2571-MSM1-P1
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15920078
  • 485.00€ / 100µg
Expédition estimée: 11-07-2024
Spécification
GAD1 / GAD67 (GABAergic Neuronal Marker)
Cytoplasmic
GAD1/2391
Unconjugated
Mouse
67kDa
Protein A/G purified from Bioreactor concentrate.
RUO
2571
IgG1 κ
ELISA, Western Blot, Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant human GAD1 (GAD67) protein fragment (apprx. aa 72-135)
K-562 or HEK293 Cells. Pancreas
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.