missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GARS Recombinant Rabbit Monoclonal Antibody (23GB5515), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA553892
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GARS is a glycyl-tRNA synthetase, one of the aminoacyl-tRNA synthetases that charge tRNAs with their cognate amino acids. This protein is an (alpha)2 dimer which belongs to the class II family of tRNA synthetases. The protein has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis.
Spécification
| GARS | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GARS1 | |
| 6.1.1.14; AIR synthase; AIRS; Ap4A synthetase; AP-4-A synthetase; Charcot-Marie-Tooth neuropathy 2D; Charcot-Marie-Tooth neuropathy, neuronal type, D; CMT2D; Diadenosine tetraphosphate synthetase; DSMAV; EC 2.1.2.29; GAR transformylase; GARS; gars1; GART; GARTF; Gena201; GENA202; glycine tRNA ligase; glycine--tRNA ligase; glycyl-tran synthetase; glycyl-tRNA synthetase; glycyl-tRNA synthetase 1; glycyl-tRNA synthetase-like protein; glyRS; HMN5; metronome; MGC47764; mrn; Nmf249; PAIS; PGFT; PRGS; RGD1559871; Sgrp23; si:dkey-276i5.1; SMAD1; storage granule protein 23; Unknown (protein for IMAGE:7254992); wu:fb02f03 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2617, 297113, 353172 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5515 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P41250, Q5I0G4, Q9CZD3 | |
| GARS, GARS1 | |
| A synthesized peptide derived from human GARS (701-739AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu