missing translation for 'onlineSavingsMsg'
Learn More

GATA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10917-2-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837623

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Transcriptional activator or repressor which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence 5′-[AT]GATA[AG]-3′ within regulatory regions of globin genes and of other genes expressed in erythroid cells.
Spécification

Spécification

GATA1
Polyclonal
Unconjugated
GATA1
ERYF1, Erythroid transcription factor, GATA 1, GATA binding factor 1, GATA1, GF 1, GF1, NF E1 DNA binding protein, NFE1, XLTT
Rabbit
Antigen Affinity Chromatography
RUO
14460, 25172, 2623
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P15976, P17679, P43429
Gata1
GATA1 Fusion Protein Ag1350
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions