missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15303-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Guanylate binding protein expression is induced by interferon. Guanylate binding proteins are characterized by their ability to specifically bind guanine nucleotides (GMP, GDP, and GTP) and are distinguished from the GTP-binding proteins by the presence of 2 binding motifs rather than 3.Spécification
GBP1 | |
Polyclonal | |
Unconjugated | |
GBP1 | |
GBP 1, GBP1, GTP binding protein 1, HuGBP 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2633 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P32455 | |
GBP1 | |
GBP1 Fusion Protein Ag7562 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |