missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDPD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16494-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The glycerophosphodiester phosphodiesterase domain containing 5 (GDPD5) protein, also known as GDE2, is a seven transmembrane, widely expressed protein that is necessary for spinal motor neuron differentiation and retinoid-induced neuronal outgrowth. Altered choline phospholipid metabolism is a hallmark of cancer, and the elevated expression of GDPD5 correlates with maligt choline phospholipid metabolite profiles in human breast cancer.Spécification
GDPD5 | |
Polyclonal | |
Unconjugated | |
GDPD5 | |
GDE2, GDPD5, PP1665, PP9363, UNQ1850/PRO3580 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
233552, 499211, 81544 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q640M6, Q8WTR4 | |
Gdpd5 | |
GDPD5 Fusion Protein Ag9622 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |