missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GGPS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14944-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene is a member of the prenyltransferase family and encodes a protein with geranylgeranyl diphosphate (GGPP) synthase activity. The enzyme catalyzes the synthesis of GGPP from farnesyl diphosphate and isopentenyl diphosphate. GGPP is an important molecule responsible for the C20-prenylation of proteins and for the regulation of a nuclear hormone receptor. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Spécification
GGPS1 | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95749, Q6F596, Q9WTN0 | |
GGPP synthase, GGPPS, GGPPS1, GGPPSase, GGPS1 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
14593, 291211, 9453 |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
GGPS1 | |
Liquid | |
Ggps1 | |
GGPS1 Fusion Protein Ag6745 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Human, Rat |