missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycophorin A / CD235a Rabbit, Clone: GYPA/1725R, enQuireBio™
Rabbit Monoclonal Antibody
Marque: EnquireBio 2993-RBM5-P1
Informations supplémentaires : Poids : 0.01000kg
Spécification
Glycophorin A / CD235a | |
Cell surface | |
GYPA/1725R | |
Unconjugated | |
NP_002091 | |
Rabbit | |
39kDa | |
Protein A/G Purified | |
RUO | |
2994 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
GYPA | |
Recombinant human full-length Glycophorin A protein | |
Erythrocytes in any human tissues | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |