missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glypican-3 Mouse, Clone: rGPC3/863, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 2719-MSM6-P1
Informations supplémentaires : Poids : 0.01000kg
Spécification
Glypican-3 | |
Cytoplasmic | |
rGPC3/863 | |
Unconjugated | |
NP_004475 | |
Mouse | |
67kDa | |
Protein A/G Purified | |
RUO | |
2719 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
GPC3 | |
Recombinant full-length human GPC3 protein | |
293T cells or Hepatocellular carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |